Methyl-4-amino-3-(benzyloxy)-5-iodobenzenecarboxylate
Supplier: Apollo Scientific
Synonyms:
Methyl 4-amino-3-(benzyloxy)-5-iodobenzoate
OR54324-1G
OR54324-500MG
APOSOR54324-1GEA
321
EUR
APOSOR54324-1G
APOSOR54324-500MG
Methyl-4-amino-3-(benzyloxy)-5-iodobenzenecarboxylate
Methyl-4-amino-3-(benzyloxy)-5-iodobenzenecarboxylate
Formula:
C₁₅H₁₄INO₃ MW: 383,19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12827829 CAS Number: 1186405-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...