6-Chlorouracil 98%

Supplier: Apollo Scientific
Warning

OR5447-25G
APOSOR5447-25GEA 21.9 EUR
APOSOR5447-25G
6-Chlorouracil 98%
6-Chlorouracil
Formula: C₄H₃ClN₂O₂
MW: 146,53 g/mol
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014595
CAS Number: 4270-27-3
EINECS: 224-258-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR