5-Iodocytosine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Amino-5-iodo-2-pyrimidinol, 4-Amino-5-iodopyrimidin-2(1H)-one
OR54491-5G
APOSOR54491-5GEA
68.3
EUR
APOSOR54491-5G
5-Iodocytosine
5-Iodocytosine
Formula:
C₄H₄IN₃O MW: 237 g/mol Melting Pt: 250 °C Storage Temperature: Freezer |
MDL Number:
MFCD00023162 CAS Number: 1122-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...