4-Methoxy-2-methylphenylacetonitrile
Supplier: Apollo Scientific
|
Danger
|
OR54502-1G
OR54502-5G
APOSOR54502-1GEA
72.7
EUR
APOSOR54502-1G
APOSOR54502-5G
4-Methoxy-2-methylphenylacetonitrile
4-Methoxy-2-methylphenylacetonitrile
Formula:
C₁₀H₁₁NO Boiling Pt: 291,4 °C Flash Pt: 122,6 °C Storage Temperature: Ambient |
MDL Number:
MFCD02178388 CAS Number: 262298-02-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...