6-Methoxy-2(3H)-benzoxazolone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Coixol, 6-Methoxybenzo[d]oxazol-2(3H)-one
OR54553-1G
OR54553-250MG
APOSOR54553-1GEA
830
EUR
APOSOR54553-1G
APOSOR54553-250MG
6-Methoxy-2(3H)-benzoxazolone
6-Methoxy-2(3H)-benzoxazolone
Formula:
C₈H₇NO₃ MW: 165,15 g/mol Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051726 CAS Number: 532-91-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...