3-Ethynylimidazo[1,2-b]pyridazine

Supplier: Apollo Scientific
Warning

OR54580-1G OR54580-5G
APOSOR54580-1GEA 0 EUR
APOSOR54580-1G APOSOR54580-5G
3-Ethynylimidazo[1,2-b]pyridazine
3-Ethynylimidazo[1,2-b]pyridazine

Intermediate in Ponatinib synthesis.

Formula: C₈H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD17012027
CAS Number: 943320-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR