Oxamic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-2-oxoacetic acid

OR54598-100G OR54598-25G
APOSOR54598-100GEA 351 EUR
APOSOR54598-100G APOSOR54598-25G
Oxamic acid 98%
Oxamic acid
Formula: C₂H₃NO₃
MW: 89,05 g/mol
Melting Pt: 207…210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008006
CAS Number: 471-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR