Fmoc-(S)-nipecotic acid

Supplier: Apollo Scientific
Warning

Synonyms: Fmoc-Nip-OH, (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-3-carboxylic acid

OR54626-1G OR54626-5G
APOSOR54626-1GEA 85.8 EUR
APOSOR54626-1G APOSOR54626-5G
Fmoc-(S)-nipecotic acid
Fmoc-(S)-nipecotic acid
Formula: C₂₁H₂₁NO₄
MW: 351,41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04112682
CAS Number: 193693-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR