Bis(2-pyridylmethyl)amine 97%

Supplier: Apollo Scientific
Warning

OR54632-25G OR54632-5G
APOSOR54632-25GEA 119 EUR
APOSOR54632-25G APOSOR54632-5G
Bis(2-pyridylmethyl)amine 97%
Bis(2-pyridylmethyl)amine
Formula: C₁₂H₁₃N₃
MW: 199,26 g/mol
Boiling Pt: 144 °C
Density: 1,11 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00129044
CAS Number: 1539-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR