(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Boc-Lys(Boc)-OH, Nα,Nepsilon-Di-Boc-L-lysine
OR54645-100G
OR54645-25G
APOSOR54645-100GEA
236
EUR
APOSOR54645-100G
APOSOR54645-25G
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid
Formula:
C₁₆H₃₀N₂O₆ MW: 346,4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00038515 CAS Number: 2483-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...