2-Aminopyrazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Aminopyrazine, Pyrazin-2-amine
OR5470-100G
OR5470-25G
OR5470-500G
APOSOR5470-100GEA
48.1
EUR
APOSOR5470-100G
APOSOR5470-25G
APOSOR5470-500G
2-Aminopyrazine
2-Aminopyrazine
Formula:
C₄H₅N₃ MW: 95,1 g/mol Melting Pt: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006137 CAS Number: 5049-61-6 EINECS: 225-748-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...