1-Chloro-2-naphthol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Chloronaphthalen-2-ol
OR55036-1G
OR55036-25G
OR55036-5G
APOSOR55036-1GEA
21.9
EUR
APOSOR55036-1G
APOSOR55036-25G
APOSOR55036-5G
1-Chloro-2-naphthol 97%
1-Chloro-2-naphthol
Formula:
C₁₀H₇ClO MW: 178.62 g/mol Boiling Pt: 157 °C/15.75mmHg Melting Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00997084 CAS Number: 633-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...