1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride ≥95%

Supplier: Apollo Scientific
Warning

Synonyms: 5,12-Dibromoanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone

OR55149-1G OR55149-5G OR55149-10G
APOSOR55149-1GEA 31.9 EUR
APOSOR55149-1G APOSOR55149-5G APOSOR55149-10G
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride ≥95%
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride
Formula: C₂₄H₆Br₂O₆
MW: 550,11 g/mol
Melting Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD11870875
CAS Number: 118129-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR