Octadecyltrimethoxysilane 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Trimethoxy(octadecyl)silane
OR55171-25G
OR55171-100G
APOSOR55171-25GEA
27.7
EUR
APOSOR55171-25G
APOSOR55171-100G
Octadecyltrimethoxysilane 95%
Octadecyltrimethoxysilane
Formula:
C₂₁H₄₆O₃Si MW: 374,68 g/mol Boiling Pt: 170 °C Melting Pt: 16…17 °C Density: 0,885 g/cm³ (25 °C) Flash Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043060 CAS Number: 3069-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...