1H-Imidazole-4-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
OR55181-1G
OR55181-5G
APOSOR55181-1GEA
125
EUR
APOSOR55181-1G
APOSOR55181-5G
1H-Imidazole-4-carbonitrile
1H-Imidazole-4-carbonitrile
Formula:
C₄H₃N₃ MW: 93,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00233358 CAS Number: 57090-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...