2-Pentyn-1-ol
Supplier: Apollo Scientific
|
Warning
|
OR5519-25G
OR5519-5G
OR5519-100G
APOSOR5519-25GEA
216
EUR
APOSOR5519-25G
APOSOR5519-5G
APOSOR5519-100G
2-Pentyn-1-ol
2-Pentyn-1-ol
Formula:
C₅H₈O Boiling Pt: 154 °C Density: 0,91 g/cm³ (25 °C) Flash Pt: 58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040915 CAS Number: 6261-22-9 EINECS: 228-411-2 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...