Boc-L-β-homovaline 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-Boc-L-β-leucine, Boc-β-HoVal-OH, Boc-β-HomoVal-OH, (R)-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid
OR55192-10G
OR55192-1G
OR55192-5G
APOSOR55192-10GEA
454
EUR
APOSOR55192-10G
APOSOR55192-1G
APOSOR55192-5G
Boc-L-β-homovaline 98%
Boc-L-beta-homovaline
Formula:
C₁₁H₂₁NO₄ MW: 231,292 g/mol Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076232 CAS Number: 183990-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...