1,22-Docosanedioic acid 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Docosanedioic acid
OR55199-5G
APOSOR55199-5GEA
97.3
EUR
APOSOR55199-5G
1,22-Docosanedioic acid 95+%
1,22-Docosanedioic acid
Formula:
C₂₂H₄₂O₄ MW: 370,57 g/mol Melting Pt: 119…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002806 CAS Number: 505-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...