5,6-Dimethoxynicotinic acid

Supplier: Apollo Scientific

OR55218-100MG OR55218-1G OR55218-250MG
APOSOR55218-100MGEA 199 EUR
APOSOR55218-100MG APOSOR55218-1G APOSOR55218-250MG
5,6-Dimethoxynicotinic acid
5,6-Dimethoxynicotinic acid
Formula: C₈H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD12401642
CAS Number: 76470-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR