7-Chlorofuro[2,3-c]pyridine

Supplier: Apollo Scientific
Warning

OR55233-100MG OR55233-1G OR55233-250MG
APOSOR55233-100MGEA 219 EUR
APOSOR55233-100MG APOSOR55233-1G APOSOR55233-250MG
7-Chlorofuro[2,3-c]pyridine
7-Chlorofuro[2,3-c]pyridine
Formula: C₇H₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD09834947
CAS Number: 84400-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR