3-Hexyne 99%

Supplier: Apollo Scientific
Danger

OR5524-25G OR5524-5G
APOSOR5524-25GEA 104 EUR
APOSOR5524-25G APOSOR5524-5G
3-Hexyne 99%
3-Hexyne
Formula: C₆H₁₀
MW: 82,15 g/mol
Boiling Pt: 80…82 °C
Density: 0,723 g/cm³ (25 °C)
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009381
CAS Number: 928-49-4
EINECS: 213-173-4
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR