Cinnamaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: trans-Cinnamaldehyde, 3-Phenylacrylaldehyde

OR55404-100G OR55404-500G
APOSOR55404-100GEA 50.9 EUR
APOSOR55404-100G APOSOR55404-500G
Cinnamaldehyde 98%
Cinnamaldehyde
Formula: C₉H₈O
MW: 132,16 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00007000
CAS Number: 104-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR