Cyclopropane 99%

Supplier: Apollo Scientific
Danger

OR55510-25G OR55510-5G OR55510-100G
APOSOR55510-25GEA 575 EUR
APOSOR55510-25G APOSOR55510-5G APOSOR55510-100G
Cyclopropane 99%
Cyclopropane
Formula: C₃H₆
MW: 42,08 g/mol
Boiling Pt: –33 °C
Melting Pt: –128 °C
Storage Temperature: Ambient
CAS Number: 75-19-4
UN: 1027
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR