Pyridine-2,5-dicarboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: Isocinchomeronic acid, 2,5-Pyridinedicarboxylic acid

OR55594-100G
APOSOR55594-100GEA 98.6 EUR
APOSOR55594-100G
Pyridine-2,5-dicarboxylic acid 98%
Pyridine-2,5-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167,12 g/mol
Melting Pt: 242…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00006297
CAS Number: 100-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR