2-Amino-5-bromo-4-methylpyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline

OR5570-100G OR5570-25G OR5570-500G
APOSOR5570-100GEA 60.9 EUR
APOSOR5570-100G APOSOR5570-25G APOSOR5570-500G
2-Amino-5-bromo-4-methylpyridine 98%
2-Amino-5-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD03427660
CAS Number: 98198-48-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR