1,4-Diisocyanatobutane
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Tetramethylene diisocyanate
OR55710-10G
OR55710-1G
OR55710-5G
APOSOR55710-10GEA
135
EUR
APOSOR55710-10G
APOSOR55710-1G
APOSOR55710-5G
1,4-Diisocyanatobutane
1,4-Diisocyanatobutane
- May form small amount of sediment.
Formula:
C₆H₈N₂O₂ MW: 140,14 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00075559 CAS Number: 4538-37-8 UN: 3334 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...