2-Methoxy-5-isopropylbenzaldehyde 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Isopropyl-2-methoxybenzaldehyde

OR55741-25G OR55741-100G
APOSOR55741-25GEA 183 EUR
APOSOR55741-25G APOSOR55741-100G
2-Methoxy-5-isopropylbenzaldehyde 97%
2-Methoxy-5-isopropylbenzaldehyde
Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Boiling Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD03095324
CAS Number: 85902-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR