2-Acetamidoisonicotinic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Acetylaminoisonicotinic acid

OR5582-1G OR5582-250MG
APOSOR5582-1GEA 29.2 EUR
APOSOR5582-1G APOSOR5582-250MG
2-Acetamidoisonicotinic acid 97%
2-Acetamidoisonicotinic acid
Formula: C₈H₈N₂O₃
MW: 180,1600 g/mol
Melting Pt: 286…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00233713
CAS Number: 54221-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR