2-Bromo-5-thiazolecarboxylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromothiazole-5-carboxylic acid
OR5591-25G
OR5591-5G
APOSOR5591-25GEA
219
EUR
APOSOR5591-25G
APOSOR5591-5G
2-Bromo-5-thiazolecarboxylic acid 97%
2-Bromo-5-thiazolecarboxylic acid
Formula:
C₄H₂BrNO₂S MW: 208,04 g/mol Melting Pt: 180…184 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04115730 CAS Number: 54045-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...