Ethyl thiazole-5-carboxylate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 5-thiazolecarboxylate
OR5607-5G
APOSOR5607-5GEA
55.1
EUR
APOSOR5607-5G
Ethyl thiazole-5-carboxylate 98%
Ethyl thiazole-5-carboxylate
Formula:
C₆H₇NO₂S MW: 157,1900 g/mol Boiling Pt: 77…78 °C/2mmHg Melting Pt: 14 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06205069 CAS Number: 32955-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...