4-Bromo-2-ethylbenzenesulphonyl chloride 95+%
Supplier: Apollo Scientific
|
Danger
|
OR5613-1G
OR5613-250MG
OR5613-5G
APOSOR5613-1GEA
78.3
EUR
APOSOR5613-1G
APOSOR5613-250MG
APOSOR5613-5G
4-Bromo-2-ethylbenzenesulphonyl chloride 95+%
4-Bromo-2-ethylbenzenesulphonyl chloride
Formula:
C₈H₈BrClO₂S Boiling Pt: 101…104 °C/0,3mmHg Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00204178 CAS Number: 175278-24-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...