7-Chlorooxindole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Chloroindolin-2-one
OR5622-1G
OR5622-5G
OR5622-25G
APOSOR5622-1GEA
0
EUR
APOSOR5622-1G
APOSOR5622-5G
APOSOR5622-25G
7-Chlorooxindole 98%
7-Chlorooxindole
Formula:
C₈H₆ClNO MW: 167,5923 g/mol Melting Pt: 221…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD06656001 CAS Number: 25369-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...