2-Chloro-4-iodopyridine

Supplier: Apollo Scientific
Warning

OR5643-5G OR5643-100G OR5643-25G
APOSOR5643-5GEA 21.9 EUR
APOSOR5643-5G APOSOR5643-100G APOSOR5643-25G
2-Chloro-4-iodopyridine
2-Chloro-4-iodopyridine
Formula: C₅H₃ClIN
MW: 239,44 g/mol
Melting Pt: 41…45 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861983
CAS Number: 153034-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR