2'-Chloroacetophenone 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone
OR5723-500G
APOSOR5723-500GEA
89.9
EUR
APOSOR5723-500G
2'-Chloroacetophenone 95%
2'-Chloroacetophenone
Formula:
C₈H₇ClO MW: 154,6 g/mol Boiling Pt: 227…230 °C Density: 1,19 g/cm³ (25 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000560 CAS Number: 2142-68-9 EINECS: 218-397-6 UN: 3416 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...