1-Methylpiperidine-4-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

OR5738-5G OR5738-25G
APOSOR5738-5GEA 29.2 EUR
APOSOR5738-5G APOSOR5738-25G
1-Methylpiperidine-4-carboxylic acid 97%
1-Methylpiperidine-4-carboxylic acid
Formula: C₇H₁₃NO₂
MW: 143,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659473
CAS Number: 68947-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR