4-Benzylcarbamoyl-3-chlorophenylboronic acid 98%

Supplier: Apollo Scientific

Synonyms: (4-(Benzylcarbamoyl)-3-chlorophenyl)boronic acid, 3-Chloro-4-(N-benzylcarbamoyl)phenylboronic acid

OR5775-1G OR5775-5G
APOSOR5775-1GEA 144 EUR
APOSOR5775-1G APOSOR5775-5G
4-Benzylcarbamoyl-3-chlorophenylboronic acid 98%
4-Benzylcarbamoyl-3-chlorophenylboronic acid
Formula: C₁₄H₁₃BClNO₃
MW: 289,5200 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD07363769
CAS Number: 850589-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR