2-Formyl-4-thiopheneboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Formylthiophene-4-boronic acid
OR5780-1G
OR5780-5G
APOSOR5780-1GEA
80
EUR
APOSOR5780-1G
APOSOR5780-5G
2-Formyl-4-thiopheneboronic acid 98%
2-Formyl-4-thiopheneboronic acid
Formula:
C₅H₅BO₃S MW: 155,97 g/mol Melting Pt: 270…272 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03002365 CAS Number: 175592-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...