1-(3-Bromophenylsulphonyl)pyrrolidine 98%

Supplier: Apollo Scientific

Synonyms: 1-((3-Bromophenyl)sulphonyl)pyrrolidine

OR5806-25G OR5806-5G
APOSOR5806-25GEA 192 EUR
APOSOR5806-25G APOSOR5806-5G
1-(3-Bromophenylsulphonyl)pyrrolidine 98%
1-(3-Bromophenylsulphonyl)pyrrolidine
Formula: C₁₀H₁₂BrNO₂S
MW: 290,1768 g/mol
Melting Pt: 70…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363829
CAS Number: 214210-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR