2-Amino-6-chloro-3-nitropyridine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine
OR5822-25G
OR5822-100G
OR5822-5G
APOSOR5822-25GEA
21.9
EUR
APOSOR5822-25G
APOSOR5822-100G
APOSOR5822-5G
2-Amino-6-chloro-3-nitropyridine 98%
2-Amino-6-chloro-3-nitropyridine
Formula:
C₅H₄ClN₃O₂ MW: 173,56 g/mol Melting Pt: 194…196 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00044350 CAS Number: 27048-04-0 EINECS: 248-188-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...