3-Nitroanthranilic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-3-nitrobenzoic acid

OR5824-100G OR5824-25G OR5824-500G
APOSOR5824-100GEA 79.9 EUR
APOSOR5824-100G APOSOR5824-25G APOSOR5824-500G
3-Nitroanthranilic acid
3-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
Melting Pt: 207…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00024261
CAS Number: 606-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR