3-Amino-2-methoxypyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methoxypyridin-3-amine, 2-Methoxy-3-pyridylamine

OR5833-5G OR5833-1G OR5833-25G
APOSOR5833-5GEA 24.8 EUR
APOSOR5833-5G APOSOR5833-1G APOSOR5833-25G
3-Amino-2-methoxypyridine 98%
3-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
MW: 124,14 g/mol
Melting Pt: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00833386
CAS Number: 20265-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR