Benzoylacetonitrile 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Phenyl-3-oxopropanenitrile, 3-Oxo-3-phenylpropanenitrile
OR5834-100G
OR5834-250G
OR5834-25G
APOSOR5834-100GEA
132
EUR
APOSOR5834-100G
APOSOR5834-250G
APOSOR5834-25G
Benzoylacetonitrile 95%
Benzoylacetonitrile
Formula:
(C₆H₅)C(O)CH₂CN MW: 145,16 g/mol Boiling Pt: 159…160 °C/10mmHg Melting Pt: 82…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001942 CAS Number: 614-16-4 EINECS: 210-369-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...