4,6-Dimethylpyrimidine-5-carboxylic acid

Supplier: Apollo Scientific
Warning

OR5844-1G OR5844-250MG OR5844-5G
APOSOR5844-1GEA 109 EUR
APOSOR5844-1G APOSOR5844-250MG APOSOR5844-5G
4,6-Dimethylpyrimidine-5-carboxylic acid
4,6-Dimethylpyrimidine-5-carboxylic acid
Formula: C₇H₈N₂O₂
MW: 152,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787545
CAS Number: 157335-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR