3-Chloropyridine
Supplier: Apollo Scientific
|
Danger
|
OR5865-100G
OR5865-25G
APOSOR5865-100GEA
60.9
EUR
APOSOR5865-100G
APOSOR5865-25G
3-Chloropyridine
3-Chloropyridine
Formula:
C₅H₄ClN MW: 113,55 g/mol Boiling Pt: 147…149 °C Density: 1,205 g/cm³ (25 °C) Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006375 CAS Number: 626-60-8 UN: 2924 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...