(S)-(+)-2-Amino-3-methyl-1-butanol 99%

Supplier: Apollo Scientific
Warning

Synonyms: L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol

OR5874-100G OR5874-25G
APOSOR5874-100GEA 128 EUR
APOSOR5874-100G APOSOR5874-25G
(S)-(+)-2-Amino-3-methyl-1-butanol 99%
(S)-(+)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
Boiling Pt: 81 °C/8mmHg
Melting Pt: 30…32 °C
Density: 0,926 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064296
CAS Number: 2026-48-4
EINECS: 217-975-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR