4-Aminopyridine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Pyridin-4-amine, 4-Pyridylamine
OR5929-250G
OR5929-50G
APOSOR5929-250GEA
284
EUR
APOSOR5929-250G
APOSOR5929-50G
4-Aminopyridine 98%
4-Aminopyridine
Formula:
C₅H₆N₂ MW: 94,12 g/mol Boiling Pt: 273 °C Melting Pt: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 UN: 2671 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...