4-Bromo-1-(2-chlorophenyl)-1H-pyrazole

Supplier: Apollo Scientific
Warning

OR59348-1G OR59348-5G
APOSOR59348-1GEA 114 EUR
APOSOR59348-1G APOSOR59348-5G
4-Bromo-1-(2-chlorophenyl)-1H-pyrazole
4-Bromo-1-(2-chlorophenyl)-1H-pyrazole
Formula: C₉H₆BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD09878407
CAS Number: 957063-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR