1-Acetyl-4-chloro-1H-indazole 98%
Supplier: Apollo Scientific
|
Warning
|
OR59366-1G
OR59366-25G
OR59366-5G
APOSOR59366-1GEA
109
EUR
APOSOR59366-1G
APOSOR59366-25G
APOSOR59366-5G
1-Acetyl-4-chloro-1H-indazole 98%
1-Acetyl-4-chloro-1H-indazole
Formula:
C₉H₇ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD09878372 CAS Number: 145439-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...