1-Acetyl-5-nitro-1H-indazole

Supplier: Apollo Scientific
Warning

Synonyms: 1-(5-Nitro-1H-indazol-1-yl)ethanone

OR59368-1G OR59368-5G
APOSOR59368-1GEA 24.8 EUR
APOSOR59368-1G APOSOR59368-5G
1-Acetyl-5-nitro-1H-indazole
1-Acetyl-5-nitro-1H-indazole
Formula: C₉H₇N₃O₃
MW: 205,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261292
CAS Number: 13436-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR