(4-Chlorophenyl)acetic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

OR5937-1KG OR5937-250G OR5937-50G
APOSOR5937-1KGEA 292 EUR
APOSOR5937-1KG APOSOR5937-250G APOSOR5937-50G
(4-Chlorophenyl)acetic acid
(4-Chlorophenyl)acetic acid
Formula: (C₆H₄Cl)CH₂COOH
MW: 170,6 g/mol
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR